Learn More
Cytiva AcroPak™ 800 Capsules with Supor™ EKV Membrane
Product Code. 17114241
Click to view available options
Outlet Style:
1 to 1.5 in. Sanitary Flange
1/2 in. Hose Barb Inlet with Filling Bell on Outlet
Unit Size:
Each
Product Code. 17114241
Brand: Cytiva 12464
AcroPak capsules designed for research applications only.
  • For buffers, tissue culture media, and other biological fluids
  • Newer technology recommended for simplified scale-up
  • High filtration area and compact size are ideal for upscale trials
TRUSTED_SUSTAINABILITY
Specifications
Material (Membrane) Hydrophilic PES
Pore Size 0.2 μm
Sterility Sterile
Filtration Area 750 cm2
Outlet Style 1/2 in. Hose Barb Inlet with Filling Bell on Outlet
Quantity 1/Ea.