Learn More
Acrosin Antibody (ACR-2), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NB500-419MFV610
Product Code. 18675249
  • € 577.00 / 0.10mL
Description
Acrosin Monoclonal antibody specifically detects Acrosin in Porcine, Bovine (Negative), Canine (Negative), Human (Negative) samples. It is validated for Flow Cytometry, Flow (Intracellular)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Acrosin
Monoclonal
mFluor Violet 610 SE
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Pig, Bovine (Negative), Canine (Negative), Human (Negative)
Purified
Flow Cytometry
ACR-2
Flow Cytometry, Flow (Intracellular)
acrosin, ACRS, preproacrosin, proacrosin
Acid extracts of boar spermatozoa were subjected to hydrophobic chromatography and the pooled fraction with reactivity to N-alpha benzoylarginine-4-nitroanilide was used for immunization.
0.1 mL
Signal Transduction
49
Store at 4C in the dark.