Learn More
Acrosin Antibody (ACR-2), mFluor Violet 610 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB500-419MFV610
This item is not returnable.
View return policy
Description
Acrosin Monoclonal antibody specifically detects Acrosin in Porcine, Bovine (Negative), Canine (Negative), Human (Negative) samples. It is validated for Flow Cytometry, Flow (Intracellular)
Specifications
| Acrosin | |
| Monoclonal | |
| mFluor Violet 610 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Pig, Bovine (Negative), Canine (Negative), Human (Negative) | |
| Purified |
| Flow Cytometry | |
| ACR-2 | |
| Flow Cytometry, Flow (Intracellular) | |
| acrosin, ACRS, preproacrosin, proacrosin | |
| Acid extracts of boar spermatozoa were subjected to hydrophobic chromatography and the pooled fraction with reactivity to N-alpha benzoylarginine-4-nitroanilide was used for immunization. | |
| 0.1 mL | |
| Signal Transduction | |
| 49 | |
| Store at 4C in the dark. | |
| IgG1 |