Learn More
Invitrogen™ Acrosin Monoclonal Antibody (ACR-2)
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  Invitrogen™ MA119180
Product Code. 11534542
  • € 534.00 / 100µg
Description
This antibody reacts with various forms of porcine acrosin (55, 53, 45 and 35 kDa), a typical serine proteinase with trypsin-like specificity. Acrosin is stored in the acrosome of undamaged spermatozoa. This antibody will not cross-react with human, canine or bovine. Immunocytochemistry: Membrane permeabilization (acetone) is essential.
ACR is the major proteinase present in the acrosome of mature spermatozoa. It is a typical serine proteinase with trypsin-like specificity. It is stored in the acrosome in its precursor form, proacrosin. The active enzyme functions in the lysis of the zona pellucida, thus facilitating penetration of the sperm through the innermost glycoprotein layers of the ovum. The mRNA for proacrosin is synthesized only in the postmeiotic stages of spermatogenesis. In humans proacrosin first appears in the haploid spermatids.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Acrosin
Monoclonal
1 mg/mL
PBS with 15mM sodium azide; pH 7.4
P08001
ACR
Acid extracts of boar spermatozoa were subjected to hydrophobic chromatography and the pooled fraction with reactivity to N-alpha benzoylarginine-4-nitroanilide was used for immunization.
100 μg
Primary
Pig
Antibody
IgG1
Flow Cytometry, Western Blot, Immunocytochemistry
ACR-2
Unconjugated
ACR
53 kDa fucose-binding protein; Acr; Acro; Acrosin; Acrosin heavy chain; acrosin light and heavy chain prepropeptide; Acrosin light chain; acrosin precursor (EC 3.4.21.10); acrosin prepropeptide; ACRS; preproacrosin; proacrosin
Mouse
Protein A
RUO
397098
4°C, do not freeze