missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Acrosomal Vesicle Protein Recombinant Rabbit Monoclonal Antibody (23GB4285), Invitrogen™
GREENER_CHOICE

Product Code. p-200088529
Change view
Click to view available options
Quantity:
20 μL
100 μL
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30089926 100 μL 100µL
30087718 20 μL 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30089926 Supplier Invitrogen™ Supplier No. MA552674

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

This gene encodes a testis-specific, differentiation antigen, acrosomal vesicle protein 1, that arises within the acrosomal vesicle during spermatogenesis, and is associated with the acrosomal membranes and matrix of mature sperm. This gene consists of 4 exons and its alternative splicing generates multiple distinct transcripts, which encode protein isoforms ranging from 81 to 265 amino acids. The longest transcript is the most abundant, comprising 53-72% of the total acrosomal vesicle protein 1 messages; the second largest transcript comprises 15-32%; the third and the fourth largest transcripts account for 3.4-8.3% and 8.7-12.5%, respectively; and the remaining transcripts combined account for < 1% of the total acrosomal vesicle protein 1 message. It is suggested that phenomena of cryptic splicing and exon skipping occur within this gene. The acrosomal vesicle protein 1 may be involved in sperm-zona binding or penetration, and it is a potential contraceptive vaccine immunogen for humans.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Acrosomal Vesicle Protein
Applications Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB4285
Concentration 1.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene ACRV1
Gene Accession No. P26436
Gene Alias acrosomal protein SP-10; Acrosomal vesicle protein 1; Acrv1; D11S4365; Msa63; MSA-63; rSP10; Sp10; SP-10; SPACA2; sperm membrane protein; sperm protein 10
Gene Symbols ACRV1
Host Species Rabbit
Immunogen A synthesized peptide derived from human Intra Acrosomal Protein (200-265AA).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.