Learn More
Cytiva Acrylic Plastic Thermostat Jacket
Brand: Cytiva 18-0011-77
Additional Details : Weight : 0.99000kg
Specifications
Thermostat | |
Thermostat Jacket | |
600 mm | |
600 mm |
Acrylic Plastic | |
60°C | |
50 mm |