missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSBG1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 415.00 - € 624.00
Specifications
| Antigen | ACSBG1 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18405362
|
Novus Biologicals
NBP2-31990-25ul |
25 μL |
€ 415.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18172204
|
Novus Biologicals
NBP2-31990 |
0.1 mL |
€ 624.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
ACSBG1 Polyclonal specifically detects ACSBG1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ACSBG1 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human | |
| Q96GR2 | |
| 23205 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: ANVIMVDTQKQLEKILKIWKQLPHLKAVVIYKEPPPNKMANVYTMEEFMELGNEVPEEALDAIIDTQQPNQCCVLV | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Western Blot 0.4 ug/ml, Immunohistochemistry 1:200 - 1:500, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Lipid and Metabolism | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| acyl-CoA synthetase bubblegum family member 1EC 6.2.1.3, BG1, BGMBG, bubblegum, FLJ30320, hBG1, hsBG, hsBGM, KIAA0631GR-LACS, lipidosin, long-chain-fatty-acid--CoA ligase ACSBG1, LPD, MGC14352, very long-chain acyl-CoA synthetase | |
| ACSBG1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title