missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSBG1 Antibody (OTI1D8), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-02554
This item is not returnable.
View return policy
Description
ACSBG1 Monoclonal specifically detects ACSBG1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ACSBG1 | |
| Monoclonal | |
| 0.74 mg/mL | |
| Western Blot 1:2000, Immunohistochemistry 1:50, Immunocytochemistry/Immunofluorescence 1:100, Immunohistochemistry-Paraffin 1:50 | |
| ACSBG1 | |
| Full length human recombinant protein of human ACSBG1(NP_055977) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI1D8 | |
| Unconjugated | |
| acyl-CoA synthetase bubblegum family member 1EC 6.2.1.3, BG1, BGMBG, bubblegum, FLJ30320, hBG1, hsBG, hsBGM, KIAA0631GR-LACS, lipidosin, long-chain-fatty-acid--CoA ligase ACSBG1, LPD, MGC14352, very long-chain acyl-CoA synthetase | |
| Mouse | |
| 81.1 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 23205 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction