missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSBG2 Antibody, Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals H00081616-B01P
This item is not returnable.
View return policy
Description
ACSBG2 Polyclonal antibody specifically detects ACSBG2 in Human samples. It is validated for Western Blot
Specifications
| ACSBG2 | |
| Polyclonal | |
| PBS (pH 7.4) | |
| acyl-CoA synthetase bubblegum family member 2BRGL, BGRMGC111089, Bubblegum-related protein, DKFZp434K1635, EC 6.2.1.3, long-chain-fatty-acid--CoA ligase ACSBG2, PRTD-NY3PRTDNY3 | |
| ACSBG2 (AAH90046.1, 1 a.a. - 666 a.a.) full-length human protein. MTGTPKTQEGAKDLEVDMNKTEVTPRLWTTCRDGEVLLRLSKHGPGHETPMTIPEFFRESVNRFGTYPALASKNGKKWEILNFNQYYEACRKAAKSLIKLGLERFHGVGILGFNSAEWFITAVGAILAGGLCVGIYATNSAEACQYVITHAKVNILLVENDQQLQKILSIPQSSLEPLKAIIQYRLPMKKNNNLYSWDDFMELGRSIPDTQLEQVIESQKANQCAVLIYTSGTTGIPKGVMLSHDNITWIAGAVTKDFKLTDKHETVVSYLPLSHIAAQMMDIWVPIKIGALTYFAQADALKGTLVSTLKEVKPTVFIGVPQIWEKIHEMVKKNSAKSMGLKKKAFVWARNIGFKVNSKKMLGKYNTPVSYRMAKTLVFSKVKTSLGLDHCHSFISGTAPLNQETAEFFLSLDIPIGELYGLSESSGPHTISNQNNYRLLSCGKILTGCKNMLFQQNKDGIGEICLWGRHIFMGYLESETETTEAIDDEGWLHSGDLGQLDGLGFLYVTGHIKEILITAGGENVPPIPVETLVKKKIPIISNAMLVGDKLKFLSMLLTLKCEMNQMSGEPLDKLNFEAINFCRGLGSQASTVTEIVKQQDPLVYKAIQQGINAVNQEAMNNAQRIEKWVILEKDFSIYGGELGPMMKLKRHFVAQKYKKQIDHMYH | |
| 0.05 mg | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| Unconjugated | |
| AAH90046.1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 81616 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction