missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSF2 Antibody (18B4), Alexa Fluor™ 750, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Bio-Techne NBP2-59501AF750
This item is not returnable.
View return policy
Description
ACSF2 Monoclonal antibody specifically detects ACSF2 in Human samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| ACSF2 | |
| Monoclonal | |
| Alexa Fluor 750 | |
| ACSMW, acyl-CoA synthetase family member 2, AVYV493, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.26, mitochondrial, PPARG binding, long chain fatty acid acyl Co-A ligase like | |
| Recombinant human ACSF2 (42-615aa) purified from E. coli | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 80221 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Western Blot, ELISA, Immunofluorescence | |
| 18B4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction