missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSF2 Antibody (OTI6E4), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-72220UV
This item is not returnable.
View return policy
Description
ACSF2 Monoclonal specifically detects ACSF2 in Human samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| ACSF2 | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ACSF2 | |
| Full length recombinant protein of human ACSF2 (NP_079425) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG1 |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI6E4 | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| ACSMW, acyl-CoA synthetase family member 2, AVYV493, EC 6.2.1, EC 6.2.1.-, EC 6.2.1.26, mitochondrial, PPARG binding, long chain fatty acid acyl Co-A ligase like | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 80221.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction