Learn More
ACSF2 Mouse anti-Human, Clone: OTI6E4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF504711
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Acyl-CoA synthases catalyze the initial reaction in fatty acid metabolism, by forming a thioester with CoA. Has some preference toward medium-chain substrates. Plays a role in adipocyte differentiation.Specifications
| ACSF2 | |
| Monoclonal | |
| Unconjugated | |
| ACSF2 | |
| ACSMW, AVYV493 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 80221 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Flow Cytometry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| OTI6E4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| Q96CM8 | |
| ACSF2 | |
| Full length human recombit protein of human ACSF2 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.