missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSF3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA525803
This item is not returnable.
View return policy
Description
ACSF3 Polyclonal Antibody for Western Blot, IHC (P)
Catalyzes the initial reaction in intramitochondrial fatty acid synthesis, by activating malonate and methylmalonate, but not acetate, into their respective CoA thioester. May have some preference toward very-long-chain substrates.
Specifications
| ACSF3 | |
| Polyclonal | |
| Unconjugated | |
| ACSF3 | |
| Acsf3; Acyl-CoA synthetase family member 3; acyl-CoA synthetase family member 3, mitochondrial; BB101783; Malonate--CoA ligase ACSF3, mitochondrial; malonyl-CoA synthetase; PSEC0197 | |
| Rabbit | |
| Antigen Affinity Chromatography, Protein A | |
| RUO | |
| 197322, 257633 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.5 mg/mL | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q3URE1, Q4G176 | |
| ACSF3 | |
| KLH conjugated synthetic peptide between 413-442 amino acids from the central region of human ACSF3. | |
| 400 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction