missing translation for 'onlineSavingsMsg'
Learn More

ACSL3, Rabbit anti-Human, Polyclonal Antibody, Abnova™

Product Code. 16177667
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16177667 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16177667 Supplier Abnova Supplier No. H00002181D01P.100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Sequence: MNNHVSSKPSTMKLKHTINPILLYFIHFLISLYTILTYIPFYFFSESRQEKSNRIKAKPVNSKPDSAYRSVNSLDGLASVLYPGCDTLDKVFTYAKNKFKNKRLLGTREVLNEEDEVQPNGKIFKKVILGQYNWLSYEDVFVRAFNFGNGLQMLGQKPKTNIAIFCETRAEWMIAAQACFMYNFQLVTLYATLGGPAIVHALNETEVTNIITSKELLQTKLKDIVSLVPRLRHIITVDGKPPTWSEFPKGIIVHTMAAVEALGAKASMENQPHSKPLPSDIAVIMYTSGSTGLPKGVMISHSNIIAGITGMAERIPELGEEDVYIGYLPLAHVLELSAELVCLSHGCRIGYSSPQTLADQSSKIKKGSKGDTSMLKPTLMAAVPEIMDRIYKNVMNKVSEMSSFQRNLFILAYNYKMEQISKGRNTPLCDSFVFRKVRSLLGGNIRLLLCGGAPLSATTQRFMNICFCCPVGQGYGLTESAGAGTISEVWDYNTGRVGAPLVCCEIKLKNWEEGGYFNTDKPHPRGEILIGGQSVTMGYYKNEAKTKADFFEDENGQRWLCTGDIGEFEPDGCLKIIDRKKDLVKLQAGEYVSLGKVEAALKNLPLVDNICAYANSYHSYVIGFVVPNQKELTELARKKGLKGTWEELCNSCEMENEVLKVLSEAAISASLEKFEIPVKIRLSPEPWTPETGLVTDAFKLKRKELKTHYQADIERMYGRK

Specifications

Antigen ACSL3
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene acyl-CoA synthetase long-chain family member 3
Gene Accession No. NM_004457.3
Gene Alias ACS3/FACL3/PRO2194
Gene Symbols ACSL3
Host Species Rabbit
Immunogen ACSL3 (NP_004448.2, 1 a.a. ∼ 720 a.a) full-length human protein.
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 2181
Target Species Human
Content And Storage Store at -20°C or lower. Aliquot to avoid repeated freezing and thawing.
Product Type Antibody
Form Purified
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.