missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSL5 Monoclonal Antibody (OTI3A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811897S
This item is not returnable.
View return policy
Description
ACSL5 Monoclonal antibody specifically detects ACSL5 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| ACSL5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9BQ39 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 79009 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A3 | |
| Unconjugated | |
| DDX50 | |
| 1700030F05Rik; ACS2; ACS5; Acsl5; acyl-CoA synthetase 5; acyl-CoA synthetase long chain family member 5; acyl-CoA synthetase long-chain family member 5; Arachidonate--CoA ligase; FACL5; FACL5 for fatty acid coenzyme A ligase 5; fatty acid coenzyme A ligase 5; fatty acid Coenzyme A ligase, long chain 5; fatty-acid-Coenzyme A ligase, long-chain 5; HGNC:16526; LACS 5; Long-chain acyl-CoA synthetase 5; long-chain fatty acid coenzyme A ligase 5; long-chain-fatty-acid--CoA ligase 5; UNQ633/PRO1250; zgc:92083 | |
| Human recombinant protein fragment corresponding to amino acids 1-287 of human DDX50 produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction