missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSL5 Monoclonal Antibody (OTI3A3), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA811897
This item is not returnable.
View return policy
Description
The protein encoded by this gene is an isozyme of the long-chain fatty-acid-coenzyme A ligase family. Although differing in substrate specificity, subcellular localization, and tissue distribution, all isozymes of this family convert free long-chain fatty acids into fatty acyl-CoA esters, and thereby play a key role in lipid biosynthesis and fatty acid degradation. This isozyme is highly expressed in uterus and spleen, and in trace amounts in normal brain, but has markedly increased levels in maligt gliomas. This gene functions in mediating fatty acid-induced glioma cell growth. Three transcript variants encoding two different isoforms have been found for this gene.Specifications
| ACSL5 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q9ULC5 | |
| ACSL5 | |
| Human recombit protein fragment corresponding to amino acids 104-620 of human ACSL5 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI3A3 | |
| Unconjugated | |
| ACSL5 | |
| ACS2, ACS5, FACL5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 51703 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction