missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSL5 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA542999
This item is not returnable.
View return policy
Description
Peptide sequence: ACNYVKLEDV ADMNYFTVNN EGEVCIKGTN VFKGYLKDPE KTQEALDSDG Sequence homology: Cow: 92%; Dog: 93%; Guinea Pig: 86%; Horse: 86%; Human: 100%; Mouse: 100%; Rabbit: 86%; Rat: 100%; Zebrafish: 93%
ACSL5 Polyclonal antibody specifically detects ACSL5 in Human samples. It is validated for Immunohistochemistry, Western Blot.
Specifications
| ACSL5 | |
| Polyclonal | |
| Unconjugated | |
| ACSL5 | |
| 1700030F05Rik; ACS2; ACS5; Acsl5; acyl-CoA synthetase 5; acyl-CoA synthetase long chain family member 5; acyl-CoA synthetase long-chain family member 5; Arachidonate--CoA ligase; FACL5; FACL5 for fatty acid coenzyme A ligase 5; fatty acid coenzyme A ligase 5; fatty acid Coenzyme A ligase, long chain 5; fatty-acid-Coenzyme A ligase, long-chain 5; HGNC:16526; LACS 5; Long-chain acyl-CoA synthetase 5; long-chain fatty acid coenzyme A ligase 5; long-chain-fatty-acid--CoA ligase 5; UNQ633/PRO1250; zgc:92083 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 51703 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9ULC5 | |
| ACSL5 | |
| Synthetic peptide directed towards the C-terminal of human ACSL5 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction