missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ ACSL6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 16698124
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16698124 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16698124 Supplier Invitrogen™ Supplier No. PA5101652

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total ACSL6.

FACL6 is involved in activation of long-chain fatty acids for both synthesis of cellular lipids, and degradation via beta-oxidation. It plays an important role in fatty acid metabolism in brain and the acyl-CoAs produced may be utilized exclusively for the synthesis of the brain lipid. FACL6 is expressed predominantly in erythrocyte precursors, in particular in reticulocytes, fetal blood cells derieved from fetal liver, haemopoietic stem cells from cord blood, bone marrow, and brain. Expression is low at earlier stages of erythroid development but is very high in reticulocytes. This protein is involved in myelodysplastic syndrome (MDS) with basophilia, acute myelogenous leukemia (AML) with eosinophilia, and acute eosinophilic leukemia (AEL). It is characterized by a chromosomal translocation t(5;12)(q31;p13) that involves ETV6 and ACSL6.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ACSL6
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Acsl6
Gene Accession No. P33124, Q91WC3, Q9UKU0
Gene Alias A330035H04Rik; ACS2; ACSL6; acyl-CoA synthetase long-chain family member 6; Arachidonate--CoA ligase; AW050338; FACL6; fatty acid Coenzyme A ligase, long chain 6; fatty-acid-Coenzyme A ligase, long-chain 6; KIAA0837; L LACS5; LACS; LACS 6; LACS2; LACS5; Lacsl; long fatty acyl-CoA synthetase 2; long-chain acyl-CoA synthetase 6; Long-chain-fatty-acid--CoA ligase 6; long-chain-fatty-acid--CoA ligase, brain isozyme; mKIAA0837
Gene Symbols Acsl6
Host Species Rabbit
Immunogen A synthesized peptide derived from human ACSL6(Accession Q9UKU0), corresponding to amino acid residues N498-G548.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117243, 216739, 23305
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.