missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACSM1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA532119
This item is not returnable.
View return policy
Description
Recommended positive controls: Jurkat, Raji, THP-1. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
ACSM1 has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). This protein acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy-and 2,3-or 3,4-unsaturated acids (in vitro). This protein functions as GTP-dependent lipoate-activating enzyme that generates the substrate for lipoyltransferase.
Specifications
| ACSM1 | |
| Polyclonal | |
| Unconjugated | |
| ACSM1 | |
| ACSM1; Acyl-CoA synthetase medium-chain family member 1; acyl-coenzyme A synthetase ACSM1, mitochondrial; Benzoate--CoA ligase; BUCS1; Butyrate CoA ligase; butyrate--CoA ligase 1; butyryl Coenzyme A synthetase 1; butyryl-coenzyme A synthetase 1; LAE; Lipoate-activating enzyme; MACS1; medium-chain acyl-CoA synthetase; middle-chain acyl-CoA synthetase 1; Xenobiotic/medium-chain fatty acid-CoA ligase HXM-B | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 116285 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q08AH1 | |
| ACSM1 | |
| Recombinant protein encompassing a sequence within the center region of human ACSM1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction