Learn More
Invitrogen™ ACSM4 Polyclonal Antibody
GREENER_CHOICE
Product Code. 15809393
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15809393 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 15809393 Supplier Invitrogen™ Supplier No. PA570980
Rabbit Polyclonal Antibody
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 100%.
Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4- unsaturated acids (in vitro).
TRUSTED_SUSTAINABILITY
Specifications
Antigen ACSM4
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene ACSM4
Gene Accession No. P0C7M7
Gene Alias ACSM4; acyl-CoA synthetase medium chain family member 4; acyl-CoA synthetase medium-chain family member 4; Acyl-coenzyme A synthetase ACSM4, mitochondrial; Olfactory specific medium-chain acyl CoA synthetase; Omacs; O-MACS
Gene Symbols ACSM4
Host Species Rabbit
Immunogen synthetic peptide directed towards the middle region of human ACSM4.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 341392
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG