missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSM5 Antibody (OTI1G1), Alexa Fluor™ 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71978AF488
This item is not returnable.
View return policy
Description
ACSM5 Monoclonal specifically detects ACSM5 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ACSM5 | |
| Monoclonal | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ACSM5 | |
| Full length recombinant protein of human ACSM5 (NP_060358) produced in HEK293T cells. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1G1 | |
| Western Blot, Immunocytochemistry/Immunofluorescence | |
| acyl-CoA synthetase medium-chain family member 5, acyl-coenzyme A synthetase ACSM5, mitochondrial, EC 6.2.1, EC 6.2.1.2, FLJ20581, MACS3 | |
| Mouse | |
| Affinity Purified | |
| Lipid and Metabolism | |
| 54988.0 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction