missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSM5 Antibody (OTI1G1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-01874
This item is not returnable.
View return policy
Description
ACSM5 Monoclonal specifically detects ACSM5 in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| ACSM5 | |
| Monoclonal | |
| 0.81 mg/mL | |
| Western Blot 1:2000, Immunocytochemistry/Immunofluorescence 1:100 | |
| ACSM5 | |
| Full length human recombinant protein of human ACSM5(NP_060358) produced in HEK293T cell. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG1 |
| Western Blot, Immunocytochemistry, Immunofluorescence | |
| OTI1G1 | |
| Unconjugated | |
| acyl-CoA synthetase medium-chain family member 5, acyl-coenzyme A synthetase ACSM5, mitochondrial, EC 6.2.1, EC 6.2.1.2, FLJ20581, MACS3 | |
| Mouse | |
| 64.6 kDa | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 54988 | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction