missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSM5 Monoclonal Antibody (OTI5D5), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA504514
This item is not returnable.
View return policy
Description
Has medium-chain fatty acid:CoA ligase activity with broad substrate specificity (in vitro). Acts on acids from C(4) to C(11) and on the corresponding 3-hydroxy- and 2,3- or 3,4- unsaturated acids (in vitro).Specifications
| ACSM5 | |
| Monoclonal | |
| 0.94 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q6NUN0 | |
| ACSM5 | |
| Full length human recombit protein of human ACSM5 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Western Blot | |
| OTI5D5 | |
| Unconjugated | |
| ACSM5 | |
| acyl-CoA synthetase medium chain family member 5 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 54988 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction