missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACSS1 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92202-0.1ml
This item is not returnable.
View return policy
Description
ACSS1 Polyclonal antibody specifically detects ACSS1 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| ACSS1 | |
| Polyclonal | |
| Western Blot 1:500-1:2000, Immunohistochemistry 1:50-1:200, Immunocytochemistry/ Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin | |
| ACAS2L, AceCS2, AceCS2L, Acetate--CoA ligase 2, Acetyl-CoA synthetase 2, acetyl-Coenzyme A synthetase 2 (AMP forming)-like, acetyl-coenzyme A synthetase 2-like, mitochondrial, acyl-CoA synthetase short-chain family member 1dJ568C11.3, EC 6.2.1, EC 6.2.1.1, FLJ45659, KIAA1846, MGC33843 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 340-689 of human ACSS1 (NP_115890.2). ITGHSYVVYGPLCNGATSVLFESTPVYPNAGRYWETVERLKINQFYGAPTAVRLLLKYGDAWVKKYDRSSLRTLGSVGEPINCEAWEWLHRVVGDSRCTLVDTWWQTETGGICIAPRPSEEGAEILPAMAMRPFFGIVPVLMDEKGSVVEGSNVSGALCISQAWPGMARTIYGDHQRFVDAYFKAYPGYYFTGDGAYRTEGGYYQITGRMDDVINISGHRLGTAEIEDAIADHPAVPESAVIGYPHDIKGEAAFAFIVVKDSAGDSDVVVQELKSMVATKIAKYAVPDEILVVKRLPKTRSGKVMRRLLRKIITSEAQELGDTTTLEDPSIIAEILSVYQKCKDKQAAAK | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 84532 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction