missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTH Antibody (AH26), Novus Biologicals™
Mouse Monoclonal Antibody has been used in 1 publication
€ 239.00 - € 527.00
Specifications
| Antigen | ACTH |
|---|---|
| Clone | AH26 |
| Concentration | 0.2 mg/ml |
| Dilution | Flow Cytometry 0.5-1ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml |
| Applications | Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18492732
|
Novus Biologicals
NBP2-29452-20ug |
20 ug |
€ 239.00
20µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18775033
|
Novus Biologicals
NBP2-29452 |
€ 527.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
ACTH Monoclonal specifically detects ACTH in Human, Mouse, Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| ACTH | |
| 0.2 mg/ml | |
| Flow Cytometry, ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| Unconjugated | |
| Mouse | |
| Neuroscience, Nutrient Sensing in the Brain | |
| P01189 | |
| 5443 | |
| Synthetic peptide corresponding to aa1-24 of human ACTH (Uniprot: P01189) | |
| Primary | |
| Store at 4C. |
| AH26 | |
| Flow Cytometry 0.5-1ug/million cells, ELISA, Immunocytochemistry/Immunofluorescence 1-2ug/ml, Immunohistochemistry-Paraffin 0.5-1.0ug/ml, Immunohistochemistry-Frozen 0.5-1.0ug/ml | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
| POMC | |
| IgG1 κ | |
| Protein A or G purified | |
| ACTH (same as Corticotropin) is a 39 amino acid active peptide produced by the anterior pituitary. This monoclonal antibody is specific to Synacthen (aa1-24 of ACTH); does not react with CLIP (aa17-39 of ACTH). POMC (pro-opiomelanocortin or corticotropin-lipotropin) is a 267 amino acid polypeptide hormone precursor that goes through extensive, tissue-specific posttranslational processing by convertases. POMC is cleaved into ten hormone chains named NPP, ACTH, alpha-MSH (Melanocyte Stimulating Hormone), beta-MSH, gamma-MSH, CLIP (corticotropin-like intermediary peptide), Lipotropin-beta, Lipotropin-gamma, beta-endorphin and Met-enkephalin. ACTH is also produced by cells of immune system (T-cells, B-cells, and macrophages) in response to stimuli associated with stress. Anti-ACTH is a useful marker in classification of pituitary tumors and the study of pituitary disease. It reacts with ACTH-producing cells (corticotrophs).It also may react with other tumors (e.g. some small cell carcinomas of the lung) causing paraneoplastic syndromes by secreting ACTH. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title