missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTH Antibody (SPM501), Janelia Fluor™ 549 - C-terminus, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-54325JF549
This item is not returnable.
View return policy
Description
ACTH Monoclonal antibody specifically detects ACTH in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), CyTOF
Specifications
| ACTH | |
| Monoclonal | |
| Janelia Fluor 549 | |
| ACTH, adrenocorticotropic hormone, adrenocorticotropin, alpha-melanocyte-stimulating hormone, alpha-MSH, beta-endorphin, beta-LPH, beta-melanocyte-stimulating hormone, beta-MSH, CLIP, corticotropin-like intermediary peptide, corticotropin-lipotropin, gamma-LPH, gamma-MSH, lipotropin beta, lipotropin gamma, LPH, melanotropin alpha, melanotropin beta, melanotropin gamma, met-enkephalin, MSH, NPP, POC, pro-ACTH-endorphin, proopiomelanocortin, pro-opiomelanocortin, proopiomelanocortin preproprotein | |
| Synthetic peptide corresponding to aa25-39 of human ACTH (NGAEDESAEAFPLEF) (Uniprot: P01189) | |
| 0.1 mL | |
| Neuroscience | |
| 5443 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| SPM501 | |
| 50 mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction