missing translation for 'onlineSavingsMsg'
Learn More

Actin Alpha 1 Cardiac Muscle Antibody (22D3), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18142640 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18142640 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18142640 Supplier Novus Biologicals Supplier No. NBP197725AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

Actin Alpha 1 Cardiac Muscle Monoclonal specifically detects Actin Alpha 1 Cardiac Muscle in Human, Mouse, Rat, Porcine, Bovine, Goat, Mammal, Rabbit, Xenopus, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Actin Alpha 1 Cardiac Muscle
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot
Classification Monoclonal
Clone 22D3
Conjugate Alexa Fluor 350
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias ACTC, actin, alpha, cardiac muscle 1, ASD5, CMD1R, CMH11, LVNC4
Gene Symbols ACTC1
Host Species Mouse
Immunogen Clone 22D3 is a mouse monoclonal IgG1 antibody derived by fusion of NS0 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a peptide comprising the N-terminal nonapeptide of alpha-cardiac actin with an acetylated N-terminus (Ac-DDEETTALVC-COOH) coupled to keyhole limpet hemocyanin through the cysteine residue.
Purification Method Protein A or G purified
Quantity 0.1 mL
Research Discipline Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers
Primary or Secondary Primary
Gene ID (Entrez) 70
Test Specificity Clone 22D3 is an antibody highly specific for a- cardiac actin, and does not cross react with other actin isoforms.
Target Species Human, Mouse, Rat, Pig, Bovine, Goat, Mammalia, Rabbit, Xenopus species, Zebrafish
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.