missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin Alpha 1 Cardiac Muscle Antibody (22D3), Alexa Fluor™ 647, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97725AF647
This item is not returnable.
View return policy
Description
Actin Alpha 1 Cardiac Muscle Monoclonal specifically detects Actin Alpha 1 Cardiac Muscle in Human, Mouse, Rat, Porcine, Bovine, Goat, Mammal, Rabbit, Xenopus, Zebrafish samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting.
Specifications
| Actin Alpha 1 Cardiac Muscle | |
| Monoclonal | |
| Alexa Fluor 647 | |
| ACTC, actin, alpha, cardiac muscle 1, ASD5, CMD1R, CMH11, LVNC4 | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Clone 22D3 is an antibody highly specific for a- cardiac actin, and does not cross react with other actin isoforms. | |
| Store at 4C in the dark. |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunoblot | |
| 22D3 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoblotting | |
| ACTC1 | |
| Clone 22D3 is a mouse monoclonal IgG1 antibody derived by fusion of NS0 mouse myeloma cells with spleen cells from a BALB/c mouse immunized with a peptide comprising the N-terminal nonapeptide of alpha-cardiac actin with an acetylated N-terminus (Ac-DDEETTALVC-COOH) coupled to keyhole limpet hemocyanin through the cysteine residue. | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers | |
| 70 | |
| Human, Mouse, Rat, Pig, Bovine, Goat, Mammalia, Rabbit, Xenopus species, Zebrafish | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction