missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin Antibody (ACTA1/360), mFluor Violet 450 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-14162MFV450
This item is not returnable.
View return policy
Description
Actin Monoclonal antibody specifically detects Actin in Human samples. It is validated for Flow Cytometry
Specifications
| Actin | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| ACTA1/360 | |
| Flow Cytometry | |
| ACTA, actin, alpha 1, skeletal muscle, alpha skeletal muscle, alpha skeletal muscle actin, alpha-actin-1, ASMA, CFTD, CFTDM, MPFD, NEM1, NEM2, NEM3 | |
| Recombinant human Actin protein (Uniprot: P68133) | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Stem Cell Markers | |
| 58 | |
| Store at 4C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction