missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin Gamma 1 Antibody (2A3), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-97720F
This item is not returnable.
View return policy
Description
Actin Gamma 1 Monoclonal antibody specifically detects Actin Gamma 1 in Human, Mouse, Rat, Porcine, Chicken, Rabbit, Zebrafish samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Specifications
| Actin Gamma 1 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Chicken | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 2A3 | |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen | |
| ACT, ACTB, ACTG, actin, cytoplasmic 2, actin, gamma 1, cytoskeletal gamma-actin, deafness, autosomal dominant 20; deafness, autosomal dominant 26, DFNA20, DFNA26, Gamma-actin | |
| Derived by fusion of NS0 Mouse myeloma cells with spleen cells from a BALB/c Mouse immunized with a peptide comprising the N-terminal nonapeptide of gamma-cytoplasmic actin with an acetylated N-terminus (Ac-EEEIAALVI-COOH) coupled to keyhole limpet hemocyanin through the cysteine residue. The antibody does not cross react with other actin isoforms. | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Stem Cell Markers | |
| 71 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction