missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin Gamma 1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-15258
This item is not returnable.
View return policy
Description
Actin Gamma 1 Polyclonal specifically detects Actin Gamma 1 in Mouse samples. It is validated for Western Blot.
Specifications
| Actin Gamma 1 | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| ACT, ACTB, ACTG, actin, cytoplasmic 2, actin, gamma 1, cytoskeletal gamma-actin, deafness, autosomal dominant 20; deafness, autosomal dominant 26, DFNA20, DFNA26, Gamma-actin | |
| Rabbit | |
| 42 kDa | |
| 0.1 mL | |
| Cell Biology, Cellular Markers, Cytoskeleton Markers, Stem Cell Markers | |
| 71 | |
| Mouse | |
| IgG |
| Western Blot | |
| 0.26mg/mL | |
| Western Blot 1:500-1:3000 | |
| P63261 | |
| ACTG1 | |
| Recombinant protein encompassing a sequence within the center region of human Actin Gamma 1. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Mouse. Expected to cross react based on sequence identity: Chimpanzee (100%), Xenopus laevis (100%), and Xenopus tropicalis (100%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction