Learn More
Novus Biologicals™ actin, gamma 2 Antibody, Novus Biologicals™
Chicken Polyclonal Antibody
Brand:  Novus Biologicals™ NBP3-31962
Product Code. 30067461
  • € 439.00 / 400µL
Description
actin, gamma 2 Polyclonal antibody specifically detects actin, gamma 2 in Human,Mouse,Rat samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY
Specifications
Specifications
actin, gamma 2
Polyclonal
Unconjugated
ACT, ACTA3alpha-actin 3, ACTE, actin, gamma 2, smooth muscle, enteric, actin, gamma-enteric smooth muscle, ACTL3, ACTSGactin-like protein, Alpha-actin-3, Gamma-2-actin, smooth muscle gamma actin, Smooth muscle gamma-actin
Chickens were immunized with purified actin from rabbit muscle emulsified in Freund's adjuvants
400 μL
Primary
Human, Mouse, Rat
IgY
Western Blot
10 mg/mL
Sodium phosphate (10 mM, pH 7.2) buffered isotonic saline (0.9%, w v), glycerol (50%, v v)
Chicken
Affinity purified
RUO
72