missing translation for 'onlineSavingsMsg'
Learn More
Learn More
actin, gamma 2 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-83925-0.1ML
This item is not returnable.
View return policy
Description
actin, gamma 2 Polyclonal specifically detects actin, gamma 2 in Human samples. It is validated for Western Blot.
Specifications
| actin, gamma 2 | |
| Polyclonal | |
| PBS, 2% Sucrose | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 72 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| ACT, ACTA3alpha-actin 3, ACTE, actin, gamma 2, smooth muscle, enteric, actin, gamma-enteric smooth muscle, ACTL3, ACTSGactin-like protein, Alpha-actin-3, Gamma-2-actin, smooth muscle gamma actin, Smooth muscle gamma-actin | |
| The immunogen is a synthetic peptide directed towards the C-terminal region of Human actin, gamma 2 | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction