missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin (Muscle Specific) Antibody (MSA/8949R), Alexa Fluor™ 532, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-24247AF532
This item is not returnable.
View return policy
Description
Actin (Muscle Specific) Monoclonal antibody specifically detects Actin (Muscle Specific) in Human samples. It is validated for ELISA
Specifications
| Actin (Muscle Specific) | |
| Monoclonal | |
| Alexa Fluor 532 | |
| ACTA, ACTC, actin, alpha 2, smooth muscle, aorta, actin, alpha cardiac muscle 1, actin, alpha skeletal muscle, actin, aortic smooth muscle, ACTSA, alpha-cardiac actin, ASD5, ASMA, Cell growth-inhibiting gene 46 protein, CFTD, CFTD1, CFTDM, CMD1R, CMH11, MPFD, NEM1, NEM2, NEM3, nemaline myopathy type 3 | |
| Recombinant protein corresponding to human muscle actin | |
| 0.1 mL | |
| Angiogenesis, Cancer, Cardiovascular Biology, Cell Biology, Cellular Markers, Developmental Biology, Extracellular Matrix, Signal Transduction, Stem Cell Markers | |
| 58 | |
| Store at 4°C in the dark. | |
| IgG κ |
| ELISA | |
| MSA/8949R | |
| 50 mM Sodium Borate | |
| Rabbit | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction