missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin (Muscle Specific) Antibody (SPM160), Novus Biologicals™
Mouse Monoclonal Antibody
€ 265.00 - € 474.00
Specifications
| Antigen | Actin (Muscle Specific) |
|---|---|
| Clone | SPM160 |
| Concentration | 0.2 mg/mL |
| Dilution | Flow Cytometry 1-2 ug/million cells, Immunohistochemistry -frozen 0.5-1.0ug/mL, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18467501
|
Novus Biologicals
NBP2-34351-0.1mg |
0.1 mg |
€ 474.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18409951
|
Novus Biologicals
NBP2-34351-0.02mg |
0.02 mg |
€ 265.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Actin (Muscle Specific) Monoclonal antibody specifically detects Actin (Muscle Specific) in Human, Rat, Rabbit samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
| Actin (Muscle Specific) | |
| 0.2 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Mouse | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Stem Cell Markers | |
| 10 mM PBS with 0.05% BSA | |
| ACTA, ACTC, actin, alpha 2, smooth muscle, aorta, actin, alpha cardiac muscle 1, actin, alpha skeletal muscle, actin, aortic smooth muscle, ACTSA, alpha-cardiac actin, ASD5, ASMA, Cell growth-inhibiting gene 46 protein, CFTD, CFTD1, CFTDM, CMD1R, CMH11, MPFD, NEM1, NEM2, NEM3, nemaline myopathy type 3 | |
| SDS extract of human myocardium. | |
| Primary | |
| Store at 4C. |
| SPM160 | |
| Flow Cytometry 1-2 ug/million cells, Immunohistochemistry -frozen 0.5-1.0ug/mL, Immunocytochemistry/ Immunofluorescence 1-2 ug/mL, Immunohistochemistry-Paraffin 1-2 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Rat, Rabbit | |
| P62736, P68032, P68133, P68032, P68032 | |
| 58 | |
| IgG1 κ | |
| Protein A or G purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title