missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin1 Antibody (mAb45a), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53123JF646
This item is not returnable.
View return policy
Description
Actin1 Monoclonal antibody specifically detects Actin1 in Plant samples. It is validated for Western Blot
Specifications
| Actin1 | |
| Monoclonal | |
| Janelia Fluor 646 | |
| AAC1, Actin-1 | |
| Purified Arabidopsis actin (ACT1) | |
| 0.1 mL | |
| Apoptosis, Cancer, Cellular Signaling | |
| Plant | |
| Purified |
| Western Blot | |
| mAb45a | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction