missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Actin8 Antibody (mAbGPa), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-53121PCP
This item is not returnable.
View return policy
Description
Actin8 Monoclonal antibody specifically detects Actin8 in Plant samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Actin8 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Plant | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry | |
| mAbGPa | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence | |
| ACT8, Actin-8 | |
| Purified Arabidopsis Actin8 | |
| 0.1 mL | |
| Primary | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction