missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Justrite Activated Carbon Filter
Activated Carbon Filter for Lithium Battery Cabinets
Brand: Justrite JCBRSOL000
Additional Details : Weight : 1.00000kg
Description
- Top-mounted, activated carbon filter, with pre-filter
- Easy filter replacement behind a hinged panel with key lock
Specifications
Activated Carbon Filter | |
11 cm | |
Lithium Battery Cabinets |
JCBRSOL000 | |
Steel | |
23 cm |