Learn More
Activated Charcoal, Norit™, SX ultra, U.S. Food chemicals codex, Honeywell Fluka™
Product Code. 15695760 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
250 g
1 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
250g
Product Code. 15695760
Brand: Honeywell-Fluka 53663250G
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Norit® SX ultra, from peat, corresponds U.S. Food chemicals codex (3rd Ed.), steam activated and acid washed, highly purified, powder
  • Norit is a registered trademark of Norit N.V.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7440-44-0
Molecular Formula C
Molecular Weight (g/mol) 12.011
MDL Number MFCD00133992
InChI Key OKTJSMMVPCPJKN-UHFFFAOYSA-N
Synonym graphite, activated charcoal, norit, mineral, carbon-12, carbono, graphene, acticarbone, anthrasorb, carbosieve
PubChem CID 5462310
ChEBI CHEBI:27594
IUPAC Name carbon
SMILES [C]
Specifications
Color Black
pH 6 to 9
Odor Odorless
Packaging Plastic Bottle
Quantity 250 g
UN Number NONH for all modes of transport
Beilstein 4360473
Formula Weight 12.01g/mol
Chemical Name or Material Activated Charcoal Norit™, Norit SX ultra