missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Active/Pro-Caspase 3 Recombinant Rabbit Monoclonal Antibody (23GB720), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
€ 207.00 - € 562.00
Specifications
| Antigen | Active/Pro-Caspase 3 |
|---|---|
| Clone | 23GB720 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30089130
![]() |
Invitrogen™
MA554044 |
100 μL |
€ 562.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089771
|
Invitrogen™
MA554045 |
20 μL |
€ 207.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Active/Pro-Caspase 3 Recombinant Monoclonal antibody specifically detects Active/Pro-Caspase 3 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| Active/Pro-Caspase 3 | |
| Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| P42574, P55213, P70677 | |
| 12367, 25402, 836 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB720 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| A830040C14Rik; AC-3; Apopain; Casp3; CASP-3; caspase 3; Caspase 3 apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis related cysteine protease; Caspase 3, apoptosis related cysteine protease (ICE-like cysteine protease); caspase 3, apoptosis-related cysteine peptidase; caspase 3, apoptosis-related cysteine protease; caspase-3; Caspase-3 subunit p12; Caspase-3 subunit p17; CC3; Cpp32; CPP-32; CPP32B; CPP32beta; cysteine protease CPP32; IRP; Lice; mldy; PARP cleavage protease; procaspase3; protein Yama; SCA-1; SREBP cleavage activity 1; Yama; yama protein | |
| A synthetic peptide of human active+pro Caspase 3 (1-175AA). | |
| Primary | |
| -20°C | |
| CASP3 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title