Learn More
Activin A Antibody (105), Allophycocyanin, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand:  Novus Biologicals NBP2-90669APC
Product Code. 18608260
  • € 607.00 / 0.10mL
Description
Activin A Monoclonal antibody specifically detects Activin A in Mouse samples. It is validated for ELISA