missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin B/Inhibin beta B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-16966
This item is not returnable.
View return policy
Description
Activin B/Inhibin beta B Polyclonal antibody specifically detects Activin B/Inhibin beta B in Human, Mouse, Zebrafish samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen).
Specifications
| Activin B/Inhibin beta B | |
| Polyclonal | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 10% Glycerol with 0.01% Thimerosal | |
| activin AB beta polypeptide, Activin beta-B chain, INHBB, inhibin beta B chain, inhibin, beta B, inhibin, beta B (activin AB beta polypeptide), Inhibin, beta-2 | |
| Rabbit | |
| 45 kDa | |
| 0.1 mL | |
| Cytokine Research, Stem Cell Signaling Pathway | |
| 3625 | |
| Human, Mouse, Zebrafish | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen) | |
| 1mg/mL | |
| Western Blot 1:500-1:3000, Immunohistochemistry 1:100-1:1000, Immunohistochemistry-Paraffin 1:100-1:1000, Immunohistochemistry-Frozen | |
| P09529 | |
| INHBB | |
| Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of human Activin B/Inhibin beta B. The exact sequence is proprietary. | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human. Expected to cross react based on sequence identity: Mouse (100%), Chicken (100%), Pig (100%), Rat (100%), Zebrafish (100%), Bovine (100%), Xenopus laevis (93%). | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction