missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin C/Inhibin beta C Antibody (CL6915), Novus Biologicals™
Mouse Monoclonal Antibody
€ 391.65 - € 546.00
Specifications
| Antigen | Activin C/Inhibin beta C |
|---|---|
| Clone | CL6915 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18626670
|
Novus Biologicals
NBP2-76513-25ul |
25 μL |
€ 415.00 € 391.65 / 25µL Save € 23.35 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18226719
|
Novus Biologicals
NBP2-76513 |
100 μL |
€ 578.00 € 546.00 / 100µL Save € 32.00 5% Off |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Activin C/Inhibin beta C Monoclonal specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot.Specifications
| Activin C/Inhibin beta C | |
| Western Blot | |
| Unconjugated | |
| Mouse | |
| Human | |
| inhibin beta C chain, IHBC, inhibin, beta C | |
| INHBC | |
| IgG1 | |
| Protein A purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| CL6915 | |
| Monoclonal | |
| Purified | |
| Cytokine Research | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| 3626.0 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: TPRAGGQCPACGGPTLELESQRELLLDLAKRSILDKLHLTQRPTLNRPVSRAALRTALQHLHGVPQGALLEDNREQECEI | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title