missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin C/Inhibin beta C Antibody (CL6916), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-76514-25ul
This item is not returnable.
View return policy
Description
Activin C/Inhibin beta C Monoclonal specifically detects Activin C/Inhibin beta C in Human samples. It is validated for Western Blot.
Specifications
| Activin C/Inhibin beta C | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.2, containing 40% glycerol with 0.02% sodium azide | |
| INHBC | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: TPRAGGQCPACGGPTLELESQRELLLDLAKRSILDKLHLTQRPTLNRPVSRAALRTALQHLHGVPQGALLEDNREQECEI | |
| 25 μL | |
| Cytokine Research | |
| 3626.0 | |
| Human | |
| Purified |
| Western Blot | |
| CL6916 | |
| Western Blot 1 μg/mL | |
| inhibin beta C chain, IHBC, inhibin, beta C | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction