missing translation for 'onlineSavingsMsg'
Learn More

Activin RIB/ALK-4 Antibody (S07-7H9), Novus Biologicals™

Rabbit Monoclonal Antibody

€ 421.00 - € 645.00

Specifications

Antigen Activin RIB/ALK-4
Clone S07-7H9
Concentration 0.3 mg/mL
Applications Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18695024
View Documents
Novus Biologicals
NBP3-19557-100ul
100 μg
€ 645.00
100µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18676544
View Documents
Novus Biologicals
NBP3-19557-50ul
50 μg
€ 421.00
50µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Activin RIB/ALK-4 Monoclonal antibody specifically detects Activin RIB/ALK-4 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications

Specifications

Activin RIB/ALK-4
0.3 mg/mL
Monoclonal
Purified
RUO
Human, Mouse, Rat
activin A receptor, type IB, Activin receptor type IB, Activin receptor-like kinase 4, ACTRIB, ACVRLK4ACTR-IB, ALK-4, ALK4activin A receptor, type II-like kinase 4, EC 2.7.11, EC 2.7.11.30, Serine/threonine-protein kinase receptor R2, SKR2activin receptor type-1B
A synthetic peptide of human Activin RIB/ALK-4 (Uniprot # P36896)
Primary
Store at 4°C short term. Store at -20°C long term. Avoid freeze-thaw cycles.
S07-7H9
Western Blot
Unconjugated
Rabbit
Protein Kinase
50mM Tris-Glycine(pH 7.4), 0.15M NaCl, 40% Glycerol, 0.05% BSA
91
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.