missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin RIIB Antibody (MM0059-6L11), Alexa Fluor™ 700, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-12013AF700
This item is not returnable.
View return policy
Description
Activin RIIB Monoclonal specifically detects Activin RIIB in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Activin RIIB | |
| Monoclonal | |
| Alexa Fluor 700 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| ACVR2B | |
| Human recombinant Activin RIIB (extracellular domain) | |
| 0.1 mL | |
| Primary | |
| Detects human Activin RIIB. Weak cross-reactivity was found to human Activin RIIA | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| MM0059-6L11 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| activin A receptor, type IIB, Activin receptor type IIB, activin receptor type-2B, ACTRIIB, ACTR-IIB, EC 2.7.11, EC 2.7.11.30, MGC116908 | |
| Mouse | |
| Protein G purified | |
| Protein Kinase | |
| 93 | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction