missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Activin RIIB Antibody (Oxford Brookes U. patent anti-Activin Beta-B) [Janelia Fluor™ 669], Novus Biologicals™
Human Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP3-28868JF669
This item is not returnable.
View return policy
Description
Activin RIIB Monoclonal antibody specifically detects Activin RIIB in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
Specifications
| Activin RIIB | |
| Monoclonal | |
| Janelia Fluor 669 | |
| activin A receptor, type IIB, Activin receptor type IIB, activin receptor type-2B, ActR-IIB, ACTRIIB, EC 2.7.11, EC 2.7.11.30, MGC116908 | |
| INHBB/ Activin B | |
| 0.1 mL | |
| Protein Kinase | |
| 93 | |
| Store at 4°C in the dark. | |
| IgG1 |
| Flow Cytometry, ELISA, Functional Assay | |
| Oxford Brookes U. patent anti-Activin Beta-B | |
| 50mM Sodium Borate | |
| Human | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction