missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACTL6B Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-91648
This item is not returnable.
View return policy
Description
ACTL6B Polyclonal specifically detects ACTL6B in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| ACTL6B | |
| Polyclonal | |
| Immunohistochemistry 1:200 - 1:500, Immunocytochemistry/Immunofluorescence 1 - 4 ug/ml, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| O94805 | |
| ACTL6B | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:CPKADFPTTVGLLAAEEGGGLELEGDKEKKGKIFHIDTNALHV | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 53 kDa BRG1-associated factor B, actin-like 6, actin-like 6B, Actin-related protein Baf53b, ACTL6, ArpNalpha, BAF53Bactin-like protein 6B, BRG1-associated factor 53B, hArpN alpha | |
| Rabbit | |
| 47 kDa | |
| 0.1 mL | |
| Extracellular Matrix | |
| 51412 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction