missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACTL9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555380
This item is not returnable.
View return policy
Description
Immunogen sequence: GHGVTYTVPV FQGYNLLHAT ERLDLAGNHL TAFLAEMLLQ AGLPLGQQDL DLVENIKHHY CYVASDFQKE QARPEQEYKR TLKLPDGRTV TLGKELFQC Highest antigen sequence identity to the following orthologs: Mouse - 82%, Rat - 83%.
Specifications
| ACTL9 | |
| Polyclonal | |
| Unconjugated | |
| ACTL9 | |
| 1700029I08Rik; actin like 9; actin-like 7A; actin-like 7-alpha; actin-like 9; actin-like protein 9; ACTL7C; ACTL9; AV259599; HSD21; testicular tissue protein Li 15; testicular tissue protein Li 9 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 284382 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8TC94 | |
| ACTL9 | |
| Recombinant protein corresponding to Human ACTL9. Recombinant protein control fragment (Product #RP-92491). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction