missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ACTL9 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5117695
This item is not returnable.
View return policy
Description
This antibody has been tested in direct-ELISA.
Specifications
| ACTL9 | |
| Polyclonal | |
| Unconjugated | |
| ACTL9 | |
| 1700029I08Rik; actin like 9; actin-like 7A; actin-like 7-alpha; actin-like 9; actin-like protein 9; ACTL7C; ACTL9; AV259599; HSD21; testicular tissue protein Li 15; testicular tissue protein Li 9 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 284382, 69481 | |
| -20°C or -80°C if preferred | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 3.25 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q8CG27, Q8TC94 | |
| ACTL9 | |
| Recombinant Human Actin-like protein 9 protein (1-160aa). | |
| 100 μg | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction