Learn More
Invitrogen™ ACTR10 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5100215
Product Code. 16606704
  • € 502.00 / 100µL
Description
Antibody detects endogenous levels of total ACTR10.
ACTR10is a protein coding gene. Gene Ontology (GO) annotations related to this gene include neutrophil degranulation.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
ACTR10
Polyclonal
Unconjugated
ACTR10
Act11; actin related protein 10; actin related protein 10 homolog; actin-related protein 10; actin-related protein 10 homolog; actin-related protein 10 homolog (S. cerevisiae); actin-related protein 10-like protein; actin-related protein 11; Actr10; ACTR11; Arp10; ARP10 actin related protein 10 homolog; ARP10 actin-related protein 10; ARP10 actin-related protein 10 homolog; Arp11; hARP11; hypothetical protein LOC511083; LOW QUALITY PROTEIN: actin-related protein 10; zgc:55635
Rabbit
Affinity chromatography
RUO
299121, 55860, 56444
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q9NZ32, Q9QZB7
ACTR10
A synthesized peptide derived from human ACTR10(Accession Q9NZ32), corresponding to amino acid residues I360-K410.
100 μL
Primary
Human, Mouse, Rat